Anti-LH Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX72764
89353-054EA 659.34 USD
89353-054
Anti-LH Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Luteinizing hormone

IHC, ICC (frozen or formalin–fixed paraffin-embedded (FFPE) tissue sections, cell smears) 1:100-1:500 using streptavidin-Biotin system or polymer system.

Type: Primary
Antigen: LH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR