Anti-LGR7 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX12714
89359-500EA 442.73 USD
89359-500
Anti-LGR7 Rabbit Polyclonal Antibody
Antibodies
The receptors for glycoprotein hormones are G protein-coupled, 7-transmembrane receptors (GPCRs) with large N-terminal extracellular domains. Leucine-rich repeat (LRR)-containing GPCRs (LGRs) form a subgroup of the GPCR superfamily and LGR7 is a member of this group (subfamily relaxin). LGR7 expression has been reported in adrenal, brain, colon, heart, kidney, liver, lung, ovary, placenta, prostate, salivary gland, small intestine, testis, and uterus.

IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LGR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR