Anti-Le-B Mouse Monoclonal Antibody [clone: LWB01 (same as 2-25LE)]

GTX72378
89353-012EA 282.43 USD
89353-012
Anti-Le-B Mouse Monoclonal Antibody [clone: LWB01 (same as 2-25LE)]
Mouse monoclonal [2-25LE] to Blood Group Lewis b

For IHC-P: Use at a dilution of 1:400. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: Le-B
Clonality: Monoclonal
Clone: LWB01 (same as 2-25LE)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR