Anti-LEU ENKEPHALIN Mouse Monoclonal Antibody [clone: M09201]

Supplier: GeneTex

GTX26542
89361-924EA 455.6 USD
89361-924
Anti-LEU ENKEPHALIN Mouse Monoclonal Antibody [clone: M09201]
Antibodies
Type: Primary
Antigen: LEU ENKEPHALIN
Clonality: Monoclonal
Clone: [M09201]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR