Anti-Legionella Pneumophila Mouse Monoclonal Antibody

Supplier: GeneTex
GTX28263
89352-376EA 726.31 USD
89352-376
Anti-Legionella Pneumophila Mouse Monoclonal Antibody
Antibodies
Detection of Legionella pneumophila.

Type: Primary
Antigen: Legionella Pneumophila
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR