Anti-LDLR Chicken Polyclonal Antibody

Supplier: GeneTex
GTX15870
89367-448EA 692.95 USD
89367-448
Anti-LDLR Chicken Polyclonal Antibody
Antibodies
LDL (Low Density Lipoprotein) is a cholesterol carrier lipoprotein responsible for delivery of lipids (cholesterol) from liver to tissues, mainly Apo B-100 and Apo M.

ELISA (direct): Use at a concentration of 0.5 - 1 ?g/ml. WB: Use at a dilution of 1/500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: LDLR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR