Anti-LCK Mouse Monoclonal Antibody [clone: LIME-06]

Supplier: Genetex

GTX78371
89366-616EA 367 USD
89366-616
Anti-LCK Mouse Monoclonal Antibody [clone: LIME-06]
Antibodies
Type: Primary
Antigen: LCK
Clonality: Monoclonal
Clone: LIME-06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR