Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LAMB2 Mouse Monoclonal Antibody [clone: C4]

Supplier: Genetex

GTX23289
89351-966EA 358.89 USD InStock
89351-966
Anti-LAMB2 Mouse Monoclonal Antibody [clone: C4]
Antibodies
Mouse monoclonal [C4] to Laminin S

For IP: Use Protein G - Use at a dilution of 1:400. For WB: Use at a dilution of 1:200. Not suitable for use in Immunohistochemistry. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: LAMB2
Clonality: Monoclonal
Clone: C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig, Rat, Guinea Pig, Rabbit, Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty