Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgL Mouse Monoclonal Antibody [clone: 3D12]

Supplier: Genetex

GTX21944
89351-686EA 386.6 USD InStock
89351-686
Anti-IgL Mouse Monoclonal Antibody [clone: 3D12]
Antibodies
Type: Primary
Antigen: IgL
Clonality: Monoclonal
Clone: 3D12
Conjugation: Unconjugated
Epitope: Light Chain
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty