Anti-Kcnd2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX16719
89360-230EA 599.33 USD
89360-230
Anti-Kcnd2 Rabbit Polyclonal Antibody
Antibodies
In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential. Potassium channels are encoded by numerous genes whose products can be classified into distinct subfamilies of which the most distinctively characterized is that of the Kv1 shaker class.

IF: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: 1/1000. Detects a band of approximately 80 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Kcnd2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Ferret, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR