Anti-Kcnc1 Mouse Monoclonal Antibody [clone: S16B-8]

Supplier: GeneTex
GTX41994
89363-488EA 569.34 USD
89363-488
Anti-Kcnc1 Mouse Monoclonal Antibody [clone: S16B-8]
Antibodies
Mouse monoclonal antibody [S16B-8] to KV3.1

We recommend a working dilution of 1-10ug/mL (WB), 0.1-1.0ug/mL (Perox) (IHC/ICC), 1.0-10ug/mL (IF).

Type: Primary
Antigen: Kcnc1
Clonality: Monoclonal
Clone: S16B-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR