Anti-KT3 TAG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX23859
89352-056EA 617.6 USD
89352-056
Anti-KT3 TAG Rabbit Polyclonal Antibody
Antibodies
KT3 epitope tag can be used to monitor expression of the protein products from transfected with cDNA constructs.

Type: Primary
Antigen: KT3 TAG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR