Anti-KSR Rat Monoclonal Antibody [clone: MINT/1/14]

GTX11719
89358-740EA 377.02 USD
89358-740
Anti-KSR Rat Monoclonal Antibody [clone: MINT/1/14]
KSR was initially considered to be a protein kinase acting in the Ras pathway, but has more recently been demonstrated to act as a scaffold protein, facilitating complex formation between RAF and MEK and ERK.

Type: Primary
Antigen: KSR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR