Anti-KSR Mouse Monoclonal Antibody

Supplier: GeneTex

GTX14516
89360-028EA 415.03 USD
89360-028
Anti-KSR Mouse Monoclonal Antibody
Antibodies
KSR was initially considered to be a protein kinase acting in the Ras pathway, but has more recently been demonstrated to act as a scaffold protein, facilitating complex formation between RAF and MEK and ERK.

  • ELISA: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Endogenous levels of KSR may be too low to be detected by WB. Immunoprecipitation prior to WB is recommended for detection of endogenous KSR. Use at a concentration of 4 μg/ml after using 10ug to immunoprecipitate KSR from 1mg of 3T3/A31 RIPA lysate. Predicted molecular weight: 120 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: KSR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR