Anti-KLK4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX23636
89361-364EA 518.8 USD
89361-364
Anti-KLK4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to KLKL1

WB: Use at a dilution of 1/500 - 1/1000. In Western blot, this antibody is expected to detect a band of approximately 39 kDa, which corresponds to the predicted molecular weight of KLK L1. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: KLK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR