Anti-KIF4A Goat Polyclonal Antibody

GTX23815
89367-574EA 515.46 USD
89367-574
Anti-KIF4A Goat Polyclonal Antibody
Antibodies
Goat polyclonal to KIF4A

IF: Use at a dilution of 1/50. WB: Use at a dilution of 1/100. Detects a band of approximately 140 kDa (predicted molecular weight: 140 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: KIF4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR