Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KIF17 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX11261
89357-120EA 325.26 USD InStock
89357-120
Anti-KIF17 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to KIF17

IF: Use at a dilution of 1/100, this is determined using rat pheochromocytoma PC12 cell line differentiated with NGF. WB: Use at a dilution of 1/1000, this concentration is determined using a mouse brain cytosolic fraction. Staining of KIF17 is specifically inhibited with the immunizing peptide. Predicted molecular weight: 170 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: KIF17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty