Anti-KI67 Rabbit Polyclonal Antibody

GTX73691
89353-254EA 500.24 USD
89353-254
Anti-KI67 Rabbit Polyclonal Antibody
Polyclonal Antibody to Human Ki - 67 Antigen

This antibody may be diluted to a titer of 1:25-1:50 in an ABC method. However, depending on the staining conditions employed, we suggest that final dilution should be determined by the user. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining. Not tested

Type: Primary
Antigen: KI67
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR