Anti-ECE3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106130
89354-510EA 557.66 USD
89354-510
Anti-ECE3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to KEL (KELl blood group, metallo-endopeptidase)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ECE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR