Anti-IRF5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113477
89357-574EA 557.66 USD
89357-574
Anti-IRF5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to IRF5 (interferon regulatory factor 5)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: IRF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR