Anti-IRF1 Rabbit Polyclonal Antibody

GTX37648
89363-044EA 376.71 USD
89363-044
Anti-IRF1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to IRF1

P-ELISA:1:500-1000, WB:1:100-500, IHC-P:1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: IRF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Dog, Pig, Rat, Sheep, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR