Anti-CXCL10 Mouse Monoclonal Antibody [clone: 4D5]

Supplier: GeneTex
GTX11451
89358-052EA 583.12 USD
89358-052
Anti-CXCL10 Mouse Monoclonal Antibody [clone: 4D5]
Antibodies
Interferon-gamma-inducible 10 kD protein (IP-10), is a CXC chemokine with chemoattractant properties for CD4-positive T cells and inhibits early normal and leukemic hemopoietic progenitor proliferation. IP-10 is produced by a wide variety of cell types ranging from neutrophils and monocytes to hepatocytes, endothelial cells and keratinocytes. The cytokine is reported to be involved in a scala of inflammatory pathologies such as HIV encephalitis, cutaneous T cell lymphoma, chronic hepatitis and acute anterior uveitis. Various observations strongly suggest a role for the CXC chemokines IL-8 and IP-10 in the regulation of angiogenic activity in cancer and in idiopathic pulmonary fibrosis.

Recommended Starting Dilutions:ELISA: 1 ug/ml - 2 ug/mlWestern Blotting: Max Dilution 1/1000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CXCL10
Clonality: Monoclonal
Clone: 4D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR