Anti-IVL Mouse Monoclonal Antibody [clone: SY8]

Supplier: GeneTex
GTX40202
89363-128EA 602.97 USD
89363-128
Anti-IVL Mouse Monoclonal Antibody [clone: SY8]
Antibodies
Mouse monoclonal [SY8] to Involucrin

IHC-P: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 68 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IVL
Clonality: Monoclonal
Clone: SY8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog, Monkey, Pig, Gorilla, Owl
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR