Anti-IL7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX74159
89365-396EA 831.23 USD
89365-396
Anti-IL7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IL7

Recommended Starting Dilutions: ELISA: 0.5 - 2.0 ug/ml Functional Assays: 0.05 - 0.08 ug/ml Western Blotting: 0.1 - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR