Anti-IL6 Rat Monoclonal Antibody [clone: MP5-20F3]

GTX27375
89352-258EA 341.41 USD
89352-258
Anti-IL6 Rat Monoclonal Antibody [clone: MP5-20F3]
Rat monoclonal [MP5 - 20F3] to IL6

EIA: Use at an assay dependent concentration. Inhib: Use at an assay dependent concentration. Neut: Use at an assay dependent concentration. IP: please see reference (Starnes) for details. IF/ICC: please see reference (Sander) for details. ELISA: 1-4 ug/ml. ELISPOT: 5 ug/ml Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user. Neutralisation of mouse IL-6; For neutralisation of biological activity, dilutions have to be made according to the amount of IL-6 to be inactivated. Before using in biological assays, the product must be filter sterilized and depending on the concentration to be used, dialyzed against culture medium to remove the sodium azide added. Inhibits IL-6-induced proliferation of the B9 assay. Useful for immunochemical investigations of murine IL-6 in structural studies and for quantitative/qualitative immunochemical detection methods. This antibody may be used as a coating antibody in EIA procedures.

Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR