Anti-IL6 Mouse Monoclonal Antibody [clone: AS16]

Supplier: GeneTex
GTX16164
89360-196EA 611.88 USD
89360-196
Anti-IL6 Mouse Monoclonal Antibody [clone: AS16]
Antibodies
Interleukin 6 (IL6) is a multifunctional 26 kD protein originally discovered in the medium of RNA-stimulated fibroblastoid cells.

ELISA: This antibody has been used as a solution-phase antibody at 2 ?g/ml in a two-site ELISA with unconjugated IL6 antibody. Typicallly, the ELISA has a working range of 0-10000 pg/ml and sensitivity 5 pg/ml with the appropriate standard. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR