Anti-IL6 Mouse Monoclonal Antibody [clone: AS12]

Supplier: GeneTex
GTX16169
89360-200EA 461.54 USD
89360-200
Anti-IL6 Mouse Monoclonal Antibody [clone: AS12]
Antibodies
Interleukin 6 (IL6) is a multifunctional 26 kD protein originally discovered in the medium of RNA-stimulated fibroblastoid cells.

FACS: Use 10?l of antibody (10^5-10^6 cells) to stain intracellular human IL6 expressed in monocytes and macrophages. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR