Anti-IL23A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX85737
89353-946EA 473.5 USD
89353-946
Anti-IL23A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IL23A

WB (1 - 2 µg/ml), IHC-P (2.5 µg/ml)

Type: Primary
Antigen: IL23A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR