Anti-IL2 Rat Monoclonal Antibody [clone: MQ1-17H12]

Supplier: GeneTex
GTX75131
89367-162EA 656 USD
89367-162
Anti-IL2 Rat Monoclonal Antibody [clone: MQ1-17H12]
Antibodies
Rat monoclonal antibody [MQ1-17H12] to IL2

Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR