Mouse monoclonal [70625.111] to IL18R1
I-ELISA: Use at a concentration of 0.5-1 ?g/ml, determined to detect a limit of ~6 ng/well of recombinant human IL18R1. Neut: Use at a concentration of 0.12 ?g/ml. GTX10788 will neutralize 50% of the bioactivity due to 50 ng/ml recombinant human IL18 using activated human T cells. WB: Use at a concentration of 1-2 ?g/ml, determined using human IL18R1 at 25 ng/lane under non-reducing conditions. Predicted molecular weight: 60 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: IL18R1
Clonality: Monoclonal
Clone: 70625.111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human