Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL18BP Rabbit Polyclonal Antibody

Supplier: Genetex

GTX70333
89364-924EA 325.27 USD InStock
89364-924
Anti-IL18BP Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to IL-18 Binding Protein

For WB: Use at 1:500 to 1:1,000 dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher

Type: Primary
Antigen: IL18BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty