Anti-IL15 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX27213
89362-494EA 630.94 USD
89362-494
Anti-IL15 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to IL15

Neut: Use at an assay dependant dilution. WB: Use at a dilution of 1/500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR