Anti-IL1B Mouse Monoclonal Antibody

Supplier: GeneTex
GTX16168
89360-198EA 690.48 USD
89360-198
Anti-IL1B Mouse Monoclonal Antibody
Antibodies
Interleukin 1 (IL1), originally known as lymphocyte activating factor (LAF), activates T cells and lymphocytes, which then proliferate and secrete interleukin 2. IL1 is primarily released from stimulated macrophages and monocytes, but also is released from several other cell types, and is thought to play a key role in inflammatory and immune responses. The two closely related agents, interleukin1 alpha (IL1 alpha) and interleukin1 beta (IL1 beta) bind to the same cell surface receptor, elicit nearly identical biological responses and share 25% homology in their amino acid sequence.

Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR