Anti-IKBKB Mouse Monoclonal Antibody [clone: 10A9B6]

Supplier: GeneTex
GTX12140
89359-198EA 635.55 USD
89359-198
Anti-IKBKB Mouse Monoclonal Antibody [clone: 10A9B6]
Antibodies
NFkB is associated with IkB proteins in the cell cytoplasm, which inhibit NFkB activity. The IkB kinase (IKK) phosphorylates IkB, and mediates IkB degradation and NFkB activation. IKK is a serine protein kinase. The beta subunit is part of the IKK complex.

IP: 2?g of antibody is recommended for performing immunoprecipitation. WB: Use at a concentration of 1-2 ?g/ml. Detects a band of approximately 87 kDa (predicted molecular weight: 90.7 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IKBKB
Clonality: Monoclonal
Clone: 10A9B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR