Anti-IKBKB Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX27611
89367-624EA 764.07 USD
89367-624
Anti-IKBKB Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to IKK beta

Dot: Use at an assay dependant dilution. ELISA: Use at an assay dependant dilution. IP: Use at an assay dependant dilution. WB: Use at a dilution of 1/1000. Detects a band of approximately 87 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IKBKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR