Anti-IgG1 Mouse Monoclonal Antibody

Supplier: GeneTex
GTX76634
89353-416EA 473.94 USD
89353-416
Anti-IgG1 Mouse Monoclonal Antibody
Antibodies
Mouse IgG1 isotype control

Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR