Anti-IgG Mouse Monoclonal Antibody [clone: 2A11]

Supplier: GeneTex
GTX27497
89352-276EA 425.48 USD
89352-276
Anti-IgG Mouse Monoclonal Antibody [clone: 2A11]
Mouse Monoclonal antibody [2A11] to Human IgG

ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: 2A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR