Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: GeneTex

GTX73151
89353-162EA 399.82 USD
89353-162
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
HRP-conjugated Goat anti-Rat IgG Polyclonal antibody

Recommended Starting Dilutions: ELISA: Max Dilution 1/1000 Immunohistology - Frozen: Max Dilution 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IGG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR