Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IGFBP1 Mouse Monoclonal Antibody [clone: 33627.11]

Supplier: Genetex
GTX10732
Mouse monoclonal [33627.11] to IGFBP1

ELISA: Use at a concentration of 2 μg/ml. In the ELISA capture assay, plates are coated with 100 μl/well of the capture antibody (2 μg/ml) in combination with 100 μl/well of a detection antibody (affinity-purified biotinylated polyclonal anti-human IGFBP1 antibody at 100 ng/ml). An ELISA range of 62.4 to 4000 pg/ml may be obtained. Neut: Use at a concentration of 10 - 40 μg/ml. The Neutralization Dose50 (ND50) for this antibody is 10-40 μg/ml in the presence of approximately 5 μg/ml of recombinant human IGFBP1 and approximately 6 ng/mL of recombinant human IGF1, using human MCF7 cells. WB: Use at a concentration of 1 - 2 μg/ml. Predicted molecular weight: 28 kDa. The detection limit for recombinant human IGFBP1 is approximately 0.5 ng/lane under non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IGFBP1
Clonality: Monoclonal
Clone: 33627.11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg GTX10732 89354-912 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol IGFBP1
Antigen Name Insulin-like Growth Factor Binding Protein 1
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 33627.11
Host Mouse
Isotype IgG1
Western Blot Yes
ELISA Yes
Neutralisation Yes
Antigen Synonyms IBP1, PP12, IGF-BP25, hIGFBP-1, AFBP
Gene ID 3484
Concentration 0.5mg/ml
Immunogen Recombinant full length protein (Human).
Purification Protein G purified
Storage Buffer Preservative: None Constituents: PBS
Storage Temperature Keep as concentrated solution. Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Tested Applications ELISA, Neut, WB

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty