Anti-IGF1 Mouse Monoclonal Antibody [clone: 1H7]

Supplier: GeneTex
GTX20986
89360-928EA 602.2 USD
89360-928
Anti-IGF1 Mouse Monoclonal Antibody [clone: 1H7]
Antibodies
The protein encoded by this gene is similar to insulin in function and structure and is a member of a family of proteins involved in mediating growth and development. The encoded protein is processed from a precursor, bound by a specific receptor, and secreted. Defects in this gene are a cause of insulin-like growth factor I deficiency. Several transcript variants encoding different isoforms have been found for this gene.

Dilute in PBS or medium which is identical to that used in the assay system. This antibody inhibits binding of IGF-1 and -2 to the receptor and inhibits cell growth.

Type: Primary
Antigen: IGF1
Clonality: Monoclonal
Clone: 1H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR