Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgE Mouse Monoclonal Antibody

Supplier: Genetex

GTX27378
89352-262EA 386.6 USD InStock
89352-262
Anti-IgE Mouse Monoclonal Antibody
Antibodies
Mouse Monoclonal antibody [E411] to Human IgE

ELISA: Use at a dilution of 1/1000000. Used for the detection of total IgE level and IgE antibodies in ELISA, MoAb GTX27378 can be used for coating. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IgE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty