Anti-IgA Mouse Monoclonal Antibody [clone: MARA-1] (FITC (Fluorescein Isothiocyanate))

Supplier: GeneTex
GTX75558
89365-832EA 492.72 USD
89365-832
Anti-IgA Mouse Monoclonal Antibody [clone: MARA-1] (FITC (Fluorescein Isothiocyanate))
Antibodies
FITC-conjugated Mouse anti-Rat IgA (Heavy chain) Monoclonal antibody [MARA-1]

Recommended Starting Dilutions: Flow Cytometry: Max Dilution 5 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IgA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy chain
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR