Anti-IgA Mouse Monoclonal Antibody [clone: AS2]

GTX27492
89352-272EA 405.93 USD
89352-272
Anti-IgA Mouse Monoclonal Antibody [clone: AS2]
Mouse Monoclonal antibody [AS2] to Human IgA

WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone: AS2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR