Anti-IgA Mouse Monoclonal Antibody [clone: AD3]

Supplier: GeneTex
GTX20770
89360-902EA 538.15 USD
89360-902
Anti-IgA Mouse Monoclonal Antibody [clone: AD3]
Antibodies
ELISA: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone: AD3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR