Anti-IgA Mouse Monoclonal Antibody [clone: AA5]

GTX27404
89352-268EA 405.93 USD
89352-268
Anti-IgA Mouse Monoclonal Antibody [clone: AA5]
Mouse Monoclonal antibody [AA5] to Human IgA

Tested in peptide ELISA. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone: AA5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR