Anti-IgA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24219
89352-120EA 466.59 USD
89352-120
Anti-IgA Rabbit Polyclonal Antibody
Antibodies
FITC-conjugated Rabbit Polyclonal antibody to Human IgA

IF: use at a dilution of 1:20-1:40 for 30 - 60 mins at RT. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR