Anti-ID1 Rabbit Monoclonal Antibody [clone: 195-14]

Supplier: GeneTex

GTX44599
89363-546EA 1306.55 USD
89363-546
Anti-ID1 Rabbit Monoclonal Antibody [clone: 195-14]
Antibodies
Rabbit monoclonal antibody [195-14] to Id1

Recommended Starting Dilutions: IHC-P: Use at 1/100 dilution and incubate for 2 hours at room temperature WB: Dilute 1/500 in a suitable diluent and incubate for 1-16 hours at room temp Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: ID1
Clonality: Monoclonal
Clone: 195-14
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR