Anti-ID1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX48657
89363-676EA 611.14 USD
89363-676
Anti-ID1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Id1

For WB: Use at a suggested starting dilution of 1:500-1:1,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ID1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR