Anti-IgG2 beta Goat Polyclonal Antibody

Supplier: GeneTex

GTX29164
89352-588EA 184.52 USD
89352-588
Anti-IgG2 beta Goat Polyclonal Antibody
Antibodies
This secondary antibody specifically recognizes Mouse IgG2b. It does not react with any other isotypes.

Type: Secondary
Antigen: IgG2 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR