Anti-KPNA4 Goat Polyclonal Antibody

Supplier: GeneTex
GTX26039
89361-808EA 607.87 USD
89361-808
Anti-KPNA4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to KPNA4 / IPOA3

Approx 55-60kDa band observed in Human Testis and Ovary lysates (predicted size of approx. 58kDa according to NP_002259.1). Recommended for use at 0.2-1µg/ml In paraffin embedded Human Testis, shows strong nuclear staining of some cells in the seminiferous tubules. Recommended concentration, 3µg/ml. In paraffin embedded Human Small Intestine shows pixulate staining in nuclei of enterocytes. Recommended concentration, 1-2µg/ml.

Type: Primary
Antigen: KPNA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR